Learn More
American Research Products Inc Biotinylated AntiMMAE ab
Supplier:  American Research Products Inc HDBS1192
Encompass
Biotinylated Anti-MMAE antibody [8C4] is a payload antibody produced in rabbit Available in 10 g size with 7 - 14 business days lead time Ships via gel pack Store at -20 C to -80 C for 12 months in lyophilized form After reconstitution if not intended for use within a month aliquot and store at -80 C (Avoid repeated freezing and thawing) Lyophilized proteins are shipped at ambient temperature
Catalog No. 50-003-06308