Learn More
American Research Products Inc Biotinylated human Fc gamma RIIIa / CD16a (176F) protein 250UG
Supplier:  American Research Products Inc HUGR3AFB
Encompass
The sequence of the extracellular domain of human CD16a (Gly 17-Gln 208) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.
Catalog No. NC3255822