Learn More
KACTUSBIO INC Biotinylated Human Claudin 18.2 Protein-VLP (CLD-HE1822B) . 1EA=100UG.
Supplier:  KACTUSBIO INC CLDHE1822B100UL/DUP
Encompass_Preferred
Recombinant Biotinylated Full length Human Claudin 18.2 Protein-VLP is expressed from HEK293.It contains Met1-Val261.
Catalog No. NC2956804