Learn More
Jackson Immuno Research Labs BIOTINYLATED GOAT ANTI-MOUSE
SDP
Supplier:  Jackson Immuno Research Labs 115066075
Encompass_Preferred
Biotin-SP-AffiniPure F(ab')2 Fragment Goat Anti-Mouse IgM, u Chain Specific (min X Hu,Bov,Hrs Sr Prot)
Catalog No. NC9625305
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov