Learn More
KACTUSBIO INC Biotinylated Human GPRC5D Protein-Nanodisc (GPR-HM45PB) . 1EA=100UG.
Supplier:  KACTUSBIO INC GPRHM45PB100UG
Encompass
Recombinant Biotinylated Human GPRC5D Protein-Nanodisc is expressed from HEK293 with His tag and Avi tag at the C-terminus.It contains Met1-Val345.
Catalog No. NC2956816