Learn More
KACTUSBIO INC Biotinylated Human GPRC5D Protein-VLP (GPR-HM05PB) . 1EA=100UG.
Supplier:  KACTUSBIO INC GPRHM05PB100UL
Encompass
Recombinant Biotinylated Human GPRC5D Protein-VLP is expressed from HEK293.It contains Met1-Val345.
Catalog No. NC2956815