Learn More
ACROBiosystems Biotinylated Human CLEC12A / MICL / CLL-1 Protein, Fc,Avitag™
Supplier:  ACROBiosystems CLAH82F725UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CLEC12A, Fc,Avitag™ (CLA-H82F7) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 51.9 kDa.
Catalog No. 50-205-5164