Learn More
ACROBiosystems Biotinylated Human HLA-A2402
Supplier:  ACROBiosystems HL1H82E325UG
Encompass_Preferred
Biotinylated Human HLA-A*2402B2MNY-ESO-1 (LLMWITQCF) Complex Protein (HL1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 305 (HLA-A*2402) Ile 21 - Met 119 (B2M) LLMWITQCF peptide (Accession # AAA59600.1 (HLA-A*2402) P61769 (B2M) LLMWITQCF).
Catalog No. 50-253-4786