Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LIF Protein, His,Avitag™
Supplier:  ACROBiosystems LIFH82E2200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human LIF, His,Avitag (LIF-H82E2) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Phe 202 (Accession # P15018-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 23.4 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human LIF, His,Avitag (Cat. No. LIF-H82E2) on SA-coated surface at 1 μg/mL (100 μL/well) can bind Human LIF R, Fc Tag (Cat. No. LIR-H4252) with a linear range of 1-31 ng/mL (QC tested).
Catalog No. 50-210-6699