Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LRRC15 / LIB Protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems LR5H82E425UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human LRRC15, His,Avitag (LR5-H82E4) is expressed from human 293 cells (HEK293). It contains AA Tyr 22 - Gly 538 (Accession # Q8TF66-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 61.5 kDa. The protein migrates as 70-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7301