Learn More
ACROBiosystems ACROBiosystems Biotinylated Human SIRP alpha / CD172a Protein, Mouse IgG2a Fc,Avitag™ (MALS verified)
Supplier:  ACROBiosystems SIAH82A3200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human SIRP alpha, Mouse IgG2a Fc,Avitag (SIA-H82A3) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession # P78324-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 65.9 kDa. The protein migrates as 85-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Fc Tag (Cat. No. CD7-H5256) at 2 μg/mL (100 μL/well) can bind Biotinylated Human SIRP alpha, Mouse IgG2a Fc,Avitag (Cat. No. SIA-H82A3) with a linear range of 0.6-10 ng/mL (QC tested).
Catalog No. 50-210-6224