Learn More
Vector Laboratories Biotinylated Lectin Kit I
SDP
Supplier:  Vector Laboratories BK1000
Encompass
Our lectin screening kits include seven lectins or lectin conjugates. These lectins have been selected to offer a variety of sugar specificities and are of the same high quality as the reagents offered individually. Each Kit I (BK-1000, FLK-2100, RLK-2200) consists of 1 mg of the following lectins or lectin conjugates: Con A, DBA, PNA, RCA120 ,SBA, UEA I, WGA. These bioinylated conjugates are prepared from our affinity-purified lectins and are optimally labeled with biotin. Essentially free of inactive lectin conjugates and containing no free biotin, these biotinylated lectins are ideal intermediates for examining glycoconjugates using the Biotin-Avidin/Streptavidin System. First a biotin-labeled lectin is added, followed by the VECTASTAIN ABC Reagent, Avidin D conjugate, or streptavidin derivative.
Catalog No. NC9267934