Learn More
Biotium Biotinylation Reagents Fluorescent Proteins Nucleotides Other Conjugates Biotin and Streptavidin Conjugates Biotin 1/EA
Supplier:  Biotium 00020
Encompass_Preferred
Biotin-cAMP diisopropylethylammonium salt is a biotin-conjugated cAMP analog thatcan be used to probe cAMP receptors ul liWhite lyophilized solid soluble in water Store at -20C and protect from light MW 826
Catalog No. 50-005-01528