Learn More
Electron Microscopy Sciences Aurion Biotinylated Albumin/Gold, EM-grade 10 nm 1 ML
SDP
Supplier:  Electron Microscopy Sciences 25299
Encompass_Preferred
Biotinylated albumin conjugates contain equimolar amounts of conjugated protein.
Particle size: EM-grade 10 nm
Catalog No. 50-247-09