Learn More
AirSea USA Biotransporter 2 – 1.5L Temperature Control Packaging, Class 6.2 Infectious Substances Category A and Category B UN 3373
Supplier:  AirSea USA 570
Encompass_Preferred
Biotransporter-2 Temperature Control, UN Combination Packaging is a white plastic outer case with carry handle containing an insulating polystyrene box. It is supplied as a complete combination packaging, including: 1 x 1.5L Biojar (Code 490)
Catalog No. NC2101140