Learn More
Bio X Cell Antibody, anti-mouse Thy1.2 (CD90.2), Clone 30H12, Rat IgG2b, Low Endotoxin
Supplier: Bio X Cell BE0066R025MG
The 30H12 monoclonal antibody reacts with mouse Thy1.2 (CD90.2), a 25-35 kDa GPI-anchored membrane glycoprotein found on thymocytes, mature T lymphocytes, hematopoietic stem cells, neurons, epithelial cells, and fibroblasts. This antibody is useful for the depletion of T lymphocytes and has been reported to induce Ca2+ flux in thymocytes. It is supplied purified from cell culture supernatant from an animal-free facility, formulated in PBS at pH 7.0, and contains no stabilizers or preservatives.
- Reacts with mouse Thy1.2 (CD90.2)
- Clone 30H12, rat IgG2b, κ isotype
- Suitable for *in vivo* ILC depletion, *in vivo* T cell depletion, and Western blot applications
- Purity ≥95% (SDS-PAGE)
- Endotoxin level ≤1 EU/mg (LAL assay)
- Sterile (0.2 μm filtration)
- Produced in an animal-free facility
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.