Learn More
Bioworld bioZOL™ - A animal total RNA Isolation, 100 mL
Supplier:  Bioworld 107600011
Encompass_Preferred
bio-WORLD's bioZOLTM-A RNA Isolation Kit is a single phase solution designed specifically for the isolation of pure, intact RNA from animal tissue and cells in less than 90 minutes. Kit utilizes guanidine thiocyanate and β-mercaptoethanol, two of the most potent inhibitors of RNase, to effectively disrupt cellular structure, while inactivating RNase released from membrane-bound organelles. Total RNA is free of protein and DNA and can be used for cDNA synthesis, Northern blotting and in vitro translation. The procedure eliminates the need for lengthy ethanol precipitations and cesium chloride gradients.Haz Mat fee applies
Catalog No. 50-196-858