Learn More
Medchemexpress LLC Biphenyl-4,4'-dicarboxylic acid | 787-70-2 | MFCD00002554 | 98.8% | 242.23 g·mol⁻¹ | C14H10O4 | 100 G

Supplier: Medchemexpress LLC HYW013937100G
4,4'-Bibenzoic acid is an organic biphenyl dicarboxylic acid used as a biochemical reagent and building block in organic synthesis and life-science research. Supplied as a high-purity solid, it is suitable for in vitro assays and in vivo formulation studies requiring reliable physicochemical properties.
- CAS number 787-70-2.
- Molecular formula C14H10O4; molecular weight 242.23 g·mol⁻¹.
- Purity 98.76%.
- Soluble in DMSO (6.67 mg/mL; ultrasonic and warming may be required).
- Formulation: clear solution in 10% DMSO/90% corn oil at ≥0.67 mg/mL for in vivo use.
- Recommended storage: room temperature for solid; -20°C to -80°C for solutions.
- Available in multiple package sizes up to 100 g and larger quantities.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.