Learn More
Sino Biological Anti-BirA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11582RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant E. coli BirA / Bifunctional protein BirA (Catalog#11582-E10E; P06709; Met1-Lys321). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-228-2540