Learn More
Medchemexpress LLC BirA protein, E. coli (His) | >90.0% | 50UG

Supplier: Medchemexpress LLC HYP70215450UG
BirA is a recombinant biotin ligase produced in E. coli with an N-terminal 6xHis tag. It activates biotin in the presence of ATP to form the biotinyl-5'-adenylate complex and is supplied as a stabilized, filtered solution for enzymatic applications and biochemical assays.
- Purity greater than 90% as determined by reducing SDS-PAGE.
- Predicted molecular weight ~36.6 kDa by SDS-PAGE.
- Formulated in 50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT.
- Endotoxin level less than 1 EU/μg (LAL method).
- Recommended storage at -80°C; avoid repeated freeze-thaw cycles.
- Available in a 50 μg solution size suitable for assay use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.