Learn More
Medchemexpress LLC BirA protein, E. coli (His-MBP) | >95.0% | 500 UG

Supplier: Medchemexpress LLC HYP75475500UG
BirA protein (E. coli, His-MBP) is a recombinant Escherichia coli biotin ligase supplied as a purified fusion protein with N-terminal His and MBP tags. It catalyzes site-specific biotinylation of Avi-tagged substrates in the presence of ATP and is intended for in vitro protein labeling, immobilization, and affinity capture workflows. The preparation is quality-controlled by SDS-PAGE and enzyme activity assay.
- Recombinant biotin ligase expressed in E. coli
- N-terminal His and MBP tags for purification and solubility
- High purity (>95%) as determined by reducing SDS-PAGE
- Specific activity greater than 100 pmol/min/μg by pyrophosphate assay
- Suitable for site-specific biotinylation of Avi-tagged proteins
- Stable when aliquoted and stored at -20 °C or -80 °C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.