Learn More
Medchemexpress LLC HY-15743 10mg Medchemexpress, Birabresib CAS:202590-98-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1574310MG
Encompass_Preferred
Medchemexpress, HY-15743 10mg Birabresib CAS:202590-98-5 Birabresib (OTX-015) is a potent bromodomain (BRD2/3/4) inhibitor with IC50s ranging from 92 to 112 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9164