Learn More
Sino Biological Recombinant Human TAF1 Protein (C-term, GST Tag)
Supplier:  Sino Biological T2835BG100
Encompass_Preferred
Recombinant human TAF1 (1032-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-2747