Learn More
Selleck Chemical LLC Birinapant 5mg 1260251-31-7 TL32711
SDP
Supplier:  Selleck Chemical LLC S70155MG
Encompass_Preferred
Birinapant (TL32711) is a SMAC mimetic antagonist, mostly to cIAP1 with Kd of <1 nM in a cell-free assay, less potent to XIAP. Birinapant helps to induce apoptosis in latent HIV-1-infected cells. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1143562