Learn More
Apexbio Technology LLC Birinapant (TL32711), 1260251-31-7, MFCD25976869, 10 mg
SDP
Supplier:  Apexbio Technology LLC A421910
Encompass_Preferred
MF: C42H56F2N8O6, MW: 806.94, Solubility: 40.35 mg/mL in DMSO, 46.9 mg/mL in EtOH,insoluble in H2O. Birinapant, also called TL32711, is a potent antagonist for XIAP with Kd value of 45 nM and cIAP1 with Kd value <1 nM.
Catalog No. 50-101-5093