Learn More
Being Scientific BIS-1 - 10.0 x 10.0 inch, 40~250rpm, Ambient+5℃ - 50℃
Supplier:  Being Scientific BS1511U
Encompass_Preferred
BEING BIS Series Incubated Shakers offer 3 sizes to fit your capacity requirements. Accuracy in speed, temperature, and run time are guaranteed by the PID microprocessor controller. Programmability is a feature that will allow a variety of shaking protocols to be used. A full line of accessories is available to accommodate a variety of tubes, flasks, and other containers. As always BEING makes sure that all our shakers come with the necessary features and alarms guarantying your safety when in operation. Robust in design, construction, and mechanics, the BIS series shakers will provide many years of service for your incubated shaking needs.
Catalog No. 50-002-17558