Learn More
Strem, An Ascensus Company CAS# 666826-16-0. 500mg. Bis(1,5-cyclooctadiene)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 98%. MFCD10567389
SDP
Supplier:  Strem, An Ascensus Company 770225500MG
Encompass
CAS# 666826-16-0. 500mg. Bis(1,5-cyclooctadiene)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 98%. MFCD10567389. Molecular Weight: 1271.80. Molecular Formula: Ir(C8H12)2+[B(C8H3F6)4]ae. Color/form: black xtl. Strem# 77-0225. http://www.strem.com/catalog/v/77-0225/
Catalog No. 50-901-13103