Learn More
Medchemexpress LLC Bis(2,4,6-trichlorophenyl) oxalate | 1165-91-9 | MFCD00043061 | >98.0% | 448.90 | C14H4Cl6O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0424381G
Encompass_Preferred
Bis(2,4,6-trichlorophenyl) oxalate is an aryl oxalate used as a chemiluminescence reagent and biochemical assay reagent. It is a solid (white to almost white powder or crystals), with CAS number 1165-91-9 and molecular weight 448.90. Recommended storage is at room temperature; solutions are stable for longer when stored cold.
Catalog No. 50-002-10214