Learn More
Medchemexpress LLC Bis(2,4,6-trichlorophenyl) oxalate | 1165-91-9 | MFCD00043061 | 98.0% | 448.90 g·mol⁻1 | C14H4Cl6O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0424385G
Encompass_Preferred
Bis(2,4,6-trichlorophenyl) oxalate is an aryl oxalate reagent supplied for research use. It is provided as a solid at high purity for laboratory applications and is commonly used in chemiluminescent assays and organic synthesis.
  • High purity (98.0%).
  • Available in small laboratory pack sizes including 5 g.
  • Used as a chemiluminescent reagent and in dye synthesis.
  • Solid form, typically packaged in a glass bottle.
  • CAS number 1165-91-9; molecular formula C14H4Cl6O4; molecular weight 448.90 g·mol⁻1.
Catalog No. 50-002-10162