Learn More
Medchemexpress LLC Bis(2-cyanoethyl) diisopropylphosphoramidite | 102690-88-0 | 95.0% | 271.30 | 5 G
SDP
Supplier:  Medchemexpress LLC HY241265G
Encompass_Preferred
Bis(2-cyanoethyl) diisopropylphosphoramidite is a phosphoramidite monomer used in the synthesis of oligonucleotides. It is for research use only and not sold to patients.
Catalog No. 50-004-05198