Learn More
Cayman Chemical BIs 2chloroIsopropyl ether 5mg
SDP
Supplier:  Cayman Chemical 206835 MG
Encompass_Preferred
A byproduct in the synthesis of ethylene and propylene glycol; has been found in municipal drinking water in the United States and waterways in the Netherlands where it is considered a POP; mutagenic in the S. typhimurium strains TA 1535 and TA 100
Catalog No. 50-298-2540