Learn More
Medchemexpress LLC Bis(4-aminophenyl)methanone | 611-98-3 | MFCD00038138 | 99.8% | 212.25 g/mol | C13H12N2O | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01429010G
Encompass_Preferred
4,4′-Diaminobenzophenone is an aromatic diamine building block (bis(4-aminophenyl)methanone) used primarily as an intermediate in the dye and pigment industry and in photosensitive materials. Supplied as an off-white to yellow solid with high reported purity, it is suitable for organic synthesis and research applications that require a bis(4-aminophenyl)methanone motif.
  • High reported purity (99.83%).
  • Off-white to yellow solid appearance.
  • Suitable as an intermediate in dyes, pigments, and photosensitive materials.
  • Useful building block for organic synthesis and research.
  • Molecular formula C13H12N2O; molecular weight 212.25 g/mol.
Catalog No. 50-003-04040