Learn More
Medchemexpress LLC α,α'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene-13C4 | 2731660-12-9 | >95.0% | 350.43 g/mol | C24H26O2 (13C4) | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6540801MG
Encompass_Preferred
Bisphenol P-13C4 is a 13C4-labeled α,α'-bis(4-hydroxyphenyl)-1,4-diisopropylbenzene provided as an analytical standard for research use. It is intended for use as a tracer or isotopic internal standard in quantitative analytical methods; consult the certificate of analysis for storage and handling details.
  • 13C4 isotopic label for use as a tracer and internal standard.
  • Suitable for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Compact 1 mg package suitable for analytical standards and method development.
  • High purity (>95%) reported for equivalent reference materials.
  • Certificate of analysis available for assay and storage conditions.
Catalog No. 50-002-84067