Learn More
Cayman Chemical bis 7Tacrine Neurochemicals
SDP
Supplier:  Cayman Chemical 100058365 MG
Encompass_Preferred
A potent acetylcholinesterase inhibitor (IC50 = 0.40 nM); protects against hydrogen peroxide-induced apoptosis in rat pheochromocytoma cells
Catalog No. 50-288-9348