Learn More
Alkali Scientific bis Acrylamide (N,N′ Methylenebisacrylamide), 25 G
Supplier: Alkali Scientific BIS003CP
Bis Acrylamide (N,N′ Methylenebisacrylamide) 25g powder is an essential reagent used for gel preparation in electrophoresis. When combined with acrylamide, it crosslinks the gel matrix, allowing for the effective separation of proteins and nucleic acids based on their size and charge. The 25g quantity is ideal for smaller-scale applications or for laboratories with low-volume electrophoresis needs. Bis acrylamide is a key component in many biochemical and molecular biology procedures, including protein analysis, DNA sequencing, and enzyme assays. Careful handling and proper storage are critical due to the neurotoxic potential of bis acrylamide and acrylamide in general.