Learn More
Medchemexpress LLC Bis-aminooxy-PEG4 | 98627-72-6 | 96.0% | C10H24N2O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140410100MG
Encompass_Preferred
Bis-aminooxy-PEG4 is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs (Proteolysis Targeting Chimeras), which contain two distinct ligands connected by a linker. One ligand targets an E3 ubiquitin ligase, while the other targets a protein of interest. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker for PROTAC synthesis
  • Enables selective degradation of target proteins
  • Utilizes the ubiquitin-proteasome system
  • Light yellow to yellow liquid appearance
Catalog No. 50-003-56999