Learn More
Strem, An Ascensus Company CAS# 1271-54-1. 1g. Bis(benzene)chromium, min. 97%. MFCD00015485
SDP
Supplier:  Strem, An Ascensus Company 2401301G
Encompass
CAS# 1271-54-1. 1g. Bis(benzene)chromium, min. 97%. MFCD00015485. Molecular Weight: 208.22. Molecular Formula: (C6H6)2Cr. Color/form: brown to black xtl. Strem# 24-0130. http://www.strem.com/catalog/v/24-0130/
Catalog No. 50-901-10839