Learn More
Strem, An Ascensus Company CAS# 1271-19-8. Bis(cyclopentadienyl)titanium dichloride, 99+% (Titanocene dichloride) MFCD00003723. 25G
SDP
Supplier:  Strem, An Ascensus Company 22020025G
Encompass
CAS# 1271-19-8. 25G. Bis(cyclopentadienyl)titanium dichloride, 99+% (Titanocene dichloride). MFCD00003723. Molecular Formula: C10H10Cl2Ti Molecular Weight: 249.00. Color/Form: red xtl. Stability: air sensitive, moisture sensitive. Strem#: 22-0200 www.strem.com/catalog/v3/22-0200/
Catalog No. 50-901-10728