Learn More
Strem, An Ascensus Company CAS# 1277-47-0. 5g. Bis(cyclopentadienyl)vanadium, sublimed, 95% (Vanadocene). MFCD00058735
SDP
Supplier:  Strem, An Ascensus Company 2301805G
Encompass
CAS# 1277-47-0. 5g. Bis(cyclopentadienyl)vanadium, sublimed, 95% (Vanadocene). MFCD00058735. Molecular Weight: 181.13. Molecular Formula: (C5H5)2V. Color/form: purple xtl. Strem# 23-0180. http://www.strem.com/catalog/v/23-0180/
Catalog No. 50-901-10801