Learn More
Strem, An Ascensus Company CAS# 685-83-6. 1g. Bis(diethylamino)chlorophosphine, min. 97%. MFCD00015493
SDP
Supplier:  Strem, An Ascensus Company 1500841G
Encompass
CAS# 685-83-6. 1g. Bis(diethylamino)chlorophosphine, min. 97%. MFCD00015493. Molecular Weight: 210.68. Molecular Formula: [(C2H5)2N]2PCl. Color/form: colorless to light yellow liq. (may contain trace insolubles) Strem# 15-0084. http://www.strem.com/catalog/v/15-0084/
Catalog No. 50-901-09702