Learn More
Medchemexpress LLC Bis(diphenylphospholano)ethane | 824395-67-7 | MFCD07369028 | 98.0% | 506.60 | C34H36P2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW021016100MG
Encompass_Preferred
Chiral diphenylphospholane-derived phosphine ligand used as an enantioselective catalyst in asymmetric transformations, notably asymmetric hydrogenation. The compound is supplied as a high-purity research reagent and requires inert storage to maintain stability.
  • Chiral diphenylphospholane-derived phosphine ligand.
  • Enables enantioselective catalysis such as asymmetric hydrogenation.
  • High chemical purity (98.0%).
  • Molecular formula C34H36P2, molecular weight 506.60.
  • Store under nitrogen at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-003-01395