Learn More
Strem, An Ascensus Company CAS# 114460-02-5. 1g. Bis(ethylcyclopentadienyl)magnesium, min. 98%. MFCD00151375
SDP
Supplier:  Strem, An Ascensus Company 1205101G
Encompass
CAS# 114460-02-5. 1g. Bis(ethylcyclopentadienyl)magnesium, min. 98%. MFCD00151375. Molecular Weight: 210.60. Molecular Formula: (C2H5C5H4)2Mg. Color/form: colorless to pale yellow liq. Strem# 12-0510. http://www.strem.com/catalog/v/12-0510/
Catalog No. 50-901-09482