Learn More
Strem, An Ascensus Company CAS# 58628-40-3. 5g. Bis(i-propylcyclopentadienyl)zirconium dichloride, 98%. MFCD00269855
SDP
Supplier:  Strem, An Ascensus Company 4010505G
Encompass
CAS# 58628-40-3. 5g. Bis(i-propylcyclopentadienyl)zirconium dichloride, 98%. MFCD00269855. Molecular Weight: 376.48. Molecular Formula: [(C3H7)C5H4]2ZrCl2. Color/form: off-white pwdr. Strem# 40-1050. http://www.strem.com/catalog/v/40-1050/
Catalog No. 50-901-11567