Learn More
Medchemexpress LLC Bis-Mal-Lysine-PEG4-TFP ester | 1426164-53-5 | 98.0% | C37H45F4N5O13 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY141002
Encompass_Preferred
Bis-Mal-Lysine-PEG4-TFP ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules connect a ligand for an E3 ubiquitin ligase with a ligand for a target protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system. It is intended for research use only. For optimal stability, store at 4°C, protected from light. When in solvent, store at -80°C for 6 months or -20°C for 1 month, also protected from light.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system for selective protein degradation
  • Highly soluble in DMSO (100 mg/mL)
  • Demonstrates good solubility in various in vivo protocols
Catalog No. 50-003-52689