Learn More
Medchemexpress LLC Bis-mal-lysine-peg4-tfp ester | 1426164-53-5 | 98.0% | C37H45F4N5O13 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14100250MG
Encompass_Preferred
Bis-Mal-Lysine-PEG4-TFP ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Consists of two different ligands connected by a linker
  • Ligand binds to an E3 ubiquitin ligase
  • Ligand binds to the target protein
Catalog No. 50-003-54883