Learn More
Medchemexpress LLC Bis-Mal-PEG11 | 97.25% | C38H62N4O17 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14099550MG
Encompass_Preferred
Bis-Mal-PEG11 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only and not sold to patients.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-55027