Learn More
Medchemexpress LLC Bis(maltolato)oxovanadium(IV) | 38213-69-3 | 317.15 g·mol⁻¹ | C12H10O7V | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11856710MM 1ML
Encompass_Preferred
Bis(maltolato)oxovanadium(IV) is a vanadium coordination complex used as a research reagent and pan-protein tyrosine phosphatase (PTP) inhibitor. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for biochemical and cell-based assays, and is also available as a solid in multiple milligram quantities. The compound has molecular formula C12H10O7V and molecular weight 317.15 g·mol⁻¹.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available as a solid in multiple milligram sizes for storage or custom preparation.
  • Acts as a pan-protein tyrosine phosphatase (PTP) inhibitor used in biochemical and cellular studies.
  • Molecular formula C12H10O7V and molecular weight 317.15 g·mol⁻¹.
  • Brown to black solid appearance; solution provided in DMSO.
  • Intended for research use in biochemical and cell-based assays.
Catalog No. 50-002-26297