Learn More
Medchemexpress LLC Bis(maltolato)oxovanadium(IV) | 38213-69-3 | MFCD02684072 | 99.9% | 317.15 | C12H10O7V | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1185675MG
Encompass_Preferred
Bis(maltolato)oxovanadium(IV) (BMOV) is a vanadium-based research reagent used as a pan-protein tyrosine phosphatase (PTP) inhibitor and an insulin sensitizer for biochemical and cellular studies. It is supplied as a brown to black solid with high reported purity for laboratory research.
  • Potent, reversible PTP inhibitor suitable for biochemical assays.
  • Reported high purity (99.89%) for reproducible results.
  • Available in small solid quantities and as DMSO solutions for convenience.
  • Molecular weight 317.15 and formula C12H10O7V for precise calculations.
  • Suitable for studies of insulin signaling and PTP family enzymes.
Catalog No. 50-002-26463