Learn More
Medchemexpress LLC Bis(maltolato)oxovanadium(IV) | 38213-69-3 | MFCD02684072 | 99.9% | 317.15 g/mol | C12H10O7V | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11856750MG
Encompass_Preferred
Bis(maltolato)oxovanadium(IV) is a vanadium coordination complex used in biochemical and metabolic research as an insulin-mimetic and pan-protein tyrosine phosphatase inhibitor. It is provided in solid and solution formats for laboratory studies.
  • Acts as an insulin-mimetic and pan-PTP inhibitor.
  • Available as a solid and as a DMSO solution for experimental flexibility.
  • High listed purity supports reproducible biochemical results.
  • Brown to black solid appearance for quick visual identification.
  • Recommended storage conditions preserve stability for long-term use.
Catalog No. 50-002-26295