Learn More
Medchemexpress LLC Bis-NH2-PEG2 | 929-59-9 | 99.8% | 10 G
Supplier: Medchemexpress LLC HY12883310G
Bis-NH2-PEG2 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- Peg-based PROTAC linker
- Used in the synthesis of PROTACs
- Consists of two different ligands connected by a linker
- Utilizes the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins